Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRNP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047761.1 | XP_017903250.1 | 2 - 3 | 1038 | 74 | |
XM_018047762.1 | XP_017903251.1 | 3 - 4 | 666 | 74 | ||
XM_018047763.1 | XP_017903252.1 | 2 - 3 | 493 | 74 | ||
GenBank mRNA | JT666426.1 | - | 3 - 4 | 370 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |