Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THOC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053712.1 | XP_017909201.1 | 5 - 6 | 973 | 102 | |
GenBank mRNA | JO439323.2 | - | 1 - 2 | 198 | 102 | |
KA330436.1 | - | 5 - 6 | 932 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |