Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062588.1 | XP_017918077.1 | 1 - 2 | 254 | 73 | |
GenBank mRNA | JO428051.1 | - | 3 - 4 | 1304 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |