Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062588.1 | XP_017918077.1 | 5 - 6 | 685 | 103 | |
XM_018062589.1 | XP_017918078.1 | 4 - 5 | 516 | 103 | ||
GenBank mRNA | JO428051.1 | - | 7 - 8 | 1735 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |