Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AATK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063621.1 | XP_017919110.1 | 11 - 12 | 3953 | 100 | |
XM_018063622.1 | XP_017919111.1 | 11 - 12 | 3926 | 100 | ||
XM_018063623.1 | XP_017919112.1 | 11 - 12 | 3953 | 100 | ||
XM_018063624.1 | XP_017919113.1 | 11 - 12 | 3782 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |