Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIPK2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043895.1 | XP_017899384.1 | 3 - 4 | 800 | 89 | |
GenBank mRNA | JT705322.1 | - | 4 - 5 | 626 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |