Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054095.1 | XP_017909584.1 | 6 - 7 | 2342 | 62 | |
XM_018054096.1 | XP_017909585.1 | 6 - 7 | 2343 | 62 | ||
GenBank mRNA | KA315308.1 | - | 2 - 3 | 882 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |