Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686916.2 | XP_005686973.1 | 5 - 6 | 820 | 58 | |
XM_005686917.3 | XP_005686974.1 | 5 - 6 | 824 | 58 | ||
GenBank mRNA | JO524794.2 | - | 1 - 2 | 121 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |