Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013969814.2 | XP_013825268.1 | 3 - 4 | 1757 | 94 | |
XM_018059229.1 | XP_017914718.1 | 3 - 4 | 562 | 94 | ||
XM_018059230.1 | XP_017914719.1 | 3 - 4 | 416 | 94 | ||
XM_018059232.1 | XP_017914721.1 | 3 - 4 | 566 | 94 | ||
GenBank mRNA | JO428789.2 | - | 3 - 4 | 618 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |