Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPRE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039484.1 | XP_017894973.1 | 4 - 5 | 767 | 62 | |
XM_018039485.1 | XP_017894974.1 | 5 - 6 | 792 | 62 | ||
XM_018039486.1 | XP_017894975.1 | 5 - 6 | 665 | 62 | ||
GenBank mRNA | JO393367.2 | - | 3 - 4 | 601 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |