Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697869.3 | XP_005697926.2 | 5 - 6 | 594 | 91 | |
XM_013974687.2 | XP_013830141.2 | 6 - 7 | 696 | 91 | ||
XM_013974688.2 | XP_013830142.2 | 6 - 7 | 693 | 91 | ||
XM_018039896.1 | XP_017895385.1 | 6 - 7 | 815 | 91 | ||
XM_018039897.1 | XP_017895386.1 | 3 - 4 | 2319 | 91 | ||
XM_018039899.1 | XP_017895388.1 | 4 - 5 | 457 | 91 | ||
XM_018039900.1 | XP_017895389.1 | 3 - 4 | 365 | 91 | ||
GenBank mRNA | JI861726.1 | - | 3 - 4 | 270 | 91 | |
JT691468.1 | - | 4 - 5 | 605 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |