Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058394.1 | XP_017913883.1 | 5 - 6 | 2259 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |