Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYCP2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038503.1 | XP_017893992.1 | 28 - 29 | 2459 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |