Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYCP2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038503.1 | XP_017893992.1 | 29 - 30 | 2515 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |