Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040152.1 | XP_017895641.1 | 3 - 4 | 341 | 75 | |
GenBank mRNA | JI861730.1 | - | 2 - 3 | 250 | 75 | |
JO352433.2 | - | 1 - 2 | 383 | 75 | ||
JO634399.2 | - | 1 - 2 | 391 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |