Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041196.1 | XP_017896685.1 | 3 - 4 | 1234 | 65 | |
XM_018041197.1 | XP_017896686.1 | 3 - 4 | 1987 | 65 | ||
XM_018041198.1 | XP_017896687.1 | 3 - 4 | 1727 | 65 | ||
XM_018041199.1 | XP_017896688.1 | 3 - 4 | 1717 | 65 | ||
GenBank mRNA | JO430129.2 | - | 2 - 3 | 1567 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |