Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042553.1 | XP_017898042.1 | 1 - 2 | 843 | 137 | |
XM_018042554.1 | XP_017898043.1 | 1 - 2 | 843 | 137 | ||
XM_018042556.1 | XP_017898045.1 | 1 - 2 | 843 | 137 | ||
XM_018042557.1 | XP_017898046.1 | 1 - 2 | 843 | 137 | ||
XM_018042558.1 | XP_017898047.1 | 1 - 2 | 843 | 137 | ||
GenBank mRNA | JO630634.2 | - | 1 - 2 | 723 | 140 | |
JO630635.2 | - | 1 - 2 | 720 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |