Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693517.3 | XP_005693574.1 | 2 - 3 | 993 | 121 | |
GenBank mRNA | JO579520.1 | - | 2 - 3 | 1719 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |