Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038414.1 | XP_017893903.1 | 2 | 1996 | 66 | |
GenBank mRNA | JT673928.1 | - | 4 | 1465 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |