Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699368.3 | XP_005699425.2 | 2 - 3 | 414 | 112 | |
GenBank mRNA | JO429766.2 | - | 2 - 3 | 314 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |