Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699369.3 | XP_005699426.2 | 1 - 2 | 407 | 85 | |
XM_005699370.3 | XP_005699427.2 | 1 - 2 | 373 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |