Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049580.1 | XP_017905069.1 | 3 - 4 | 482 | 86 | |
XM_018049581.1 | XP_017905070.1 | 3 - 4 | 462 | 86 | ||
GenBank mRNA | JO518901.2 | - | 2 - 3 | 264 | 86 | |
JT653964.1 | - | 5 - 6 | 509 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |