Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEURL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054850.1 | XP_017910339.1 | 3 - 4 | 937 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |