Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061830.1 | XP_017917319.1 | 3 - 4 | 1596 | 60 | |
GenBank mRNA | JO623495.2 | - | 1 - 2 | 600 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |