Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061830.1 | XP_017917319.1 | 4 - 5 | 2495 | 78 | |
GenBank mRNA | JO623491.2 | - | 1 - 2 | 519 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |