Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013970980.2 | XP_013826434.2 | 1 - 2 | 1025 | 68 | |
XM_018061820.1 | XP_017917309.1 | 1 - 2 | 1018 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |