Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C18H19orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062416.1 | XP_017917905.1 | 4 - 5 | 364 | 59 | |
XM_018062417.1 | XP_017917906.1 | 4 - 5 | 348 | 59 | ||
XM_018062418.1 | XP_017917907.1 | 4 - 5 | 364 | 59 | ||
XM_018062419.1 | XP_017917908.1 | 4 - 5 | 364 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |