Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL4I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062816.1 | XP_017918305.1 | 6 - 7 | 835 | 81 | |
XM_018062817.1 | XP_017918306.1 | 6 - 7 | 696 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |