Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064568.1 | XP_017920057.1 | 5 - 6 | 598 | 69 | |
GenBank mRNA | KP408266.1 | - | 5 - 6 | 468 | 69 | |
KP408267.1 | - | 2 - 3 | 239 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |