Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JMJD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063726.1 | XP_017919215.1 | 7 - 8 | 1449 | 60 | |
XM_018063728.1 | XP_017919217.1 | 6 - 7 | 1418 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Capra hircus Probe spans exons |