Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUZP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013975878.2 | XP_013831332.2 | 10 - 11 | 1030 | 77 | |
XM_018043589.1 | XP_017899078.1 | 10 - 11 | 962 | 77 | ||
GenBank mRNA | JO643950.2 | - | 10 - 11 | 1260 | 77 | |
JT730218.1 | - | 3 - 4 | 463 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |