Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046119.1 | XP_017901608.1 | 2 - 3 | 233 | 61 | |
GenBank mRNA | KA329038.1 | - | 1 - 2 | 672 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |