Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678534.3 | XP_005678591.1 | 2 - 3 | 242 | 90 | |
GenBank mRNA | JO535987.2 | - | 2 - 3 | 210 | 90 |
Gene Symbol: PRDX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO354061.2 | - | 745 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |