Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679333.3 | XP_005679390.1 | 3 - 4 | 332 | 69 | |
GenBank mRNA | JO376554.2 | - | 4 - 5 | 242 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |