Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005683655.3 | XP_005683712.1 | 3 - 4 | 429 | 106 | |
XM_018051947.1 | XP_017907436.1 | 4 - 5 | 669 | 106 | ||
XM_018051948.1 | XP_017907437.1 | 5 - 6 | 818 | 106 | ||
XM_018051949.1 | XP_017907438.1 | 3 - 4 | 537 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |