Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005683655.3 | XP_005683712.1 | 5 - 6 | 677 | 103 | |
XM_018051947.1 | XP_017907436.1 | 6 - 7 | 917 | 103 | ||
XM_018051948.1 | XP_017907437.1 | 7 - 8 | 1066 | 103 | ||
XM_018051949.1 | XP_017907438.1 | 5 - 6 | 785 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |