Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684236.3 | XP_005684293.1 | 1 - 2 | 174 | 81 | |
GenBank mRNA | JO580075.1 | - | 1 - 2 | 259 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |