Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKLE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051374.1 | XP_017906863.1 | 6 - 7 | 1558 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |