Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052407.1 | XP_017907896.1 | 4 - 5 | 389 | 80 | |
XM_018052408.1 | XP_017907897.1 | 4 - 5 | 869 | 80 | ||
XM_018052409.1 | XP_017907898.1 | 4 - 5 | 383 | 80 | ||
XM_018052410.1 | XP_017907899.1 | 3 - 4 | 367 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |