Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057731.1 | XP_017913220.1 | 8 - 9 | 1406 | 85 | |
XM_018057732.1 | XP_017913221.1 | 7 - 8 | 1323 | 85 | ||
XM_018057733.1 | XP_017913222.1 | 7 - 8 | 1358 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |