Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046213.1 | XP_017901702.1 | 8 - 9 | 1343 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |