Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHI3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677907.3 | XP_005677964.1 | 10 - 11 | 1247 | 72 | |
XM_018045908.1 | XP_017901397.1 | 9 - 10 | 1182 | 72 | ||
GenBank mRNA | JT686748.1 | - | 4 - 5 | 619 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |