Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHI3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677907.3 | XP_005677964.1 | 4 - 5 | 404 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |