Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHI3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677907.3 | XP_005677964.1 | 5 - 6 | 545 | 101 | |
XM_018045908.1 | XP_017901397.1 | 4 - 5 | 480 | 101 | ||
GenBank mRNA | JT686748.1 | - | 1290 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |