Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHI3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677907.3 | XP_005677964.1 | 7 - 8 | 804 | 68 | |
XM_018045908.1 | XP_017901397.1 | 6 - 7 | 739 | 68 | ||
GenBank mRNA | JT686748.1 | - | 7 - 8 | 1066 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |