Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680753.3 | XP_005680810.1 | 5 - 6 | 1049 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |