Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050371.1 | XP_017905860.1 | 4 - 5 | 1022 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |