Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOMER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685075.3 | XP_005685132.1 | 3 - 4 | 1384 | 66 | |
GenBank mRNA | JT716212.1 | - | 8 - 9 | 1190 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |