Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685254.2 | XP_005685311.1 | 1 | 914 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |