Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689440.3 | XP_005689497.2 | 2 - 3 | 190 | 80 | |
XM_005689441.3 | XP_005689498.2 | 2 - 3 | 149 | 80 | ||
XM_005689442.3 | XP_005689499.2 | 3 - 4 | 160 | 80 | ||
XM_005689443.3 | XP_005689500.2 | 2 - 3 | 700 | 80 | ||
XM_005689444.3 | XP_005689501.2 | 2 - 3 | 153 | 80 | ||
GenBank mRNA | JO424465.1 | - | 4 - 5 | 2011 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |